Isochroman 99%

Supplier: Thermo Scientific Chemicals

Synonyms: 3,4-Dihydro-1H-2-benzopyran

12254-0050 12254-0250
TS12254-0050EA 35 USD
TS12254-0050 TS12254-0250
Isochroman 99%
Isochroman
Formula: C₉H₁₀O
Boiling Pt: 75 °C (4mbar)
Density: 1.06 g/cm³
Flash Pt: 65 °C
Storage Temperature: Ambient
MDL Number: MFCD00006913
CAS Number: 493-05-0

Order Now

Specification Test Results

Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5430 to 1.5460 (20 °C, 589 nm)
GC ≥98.5 %
Titration with NaOH ≥98.5 % (On anhydrous basis)
Heavy metals (as Pb) ≤5 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR