Glyoxylic acid monohydrate 98%, pure

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Formylformic acid monohydrateOxoacetic acid monohydrate2-Oxoethanoic acid monohydrate2,2-Dihydroxyacetic acid monohydrate

12017-5000 12017-0250 12017-1000
TS12017-5000EA 440 USD
TS12017-5000 TS12017-0250 TS12017-1000
Glyoxylic acid monohydrate 98%, pure
Glyoxylic acid monohydrate
Formula: C₂H₂O₃·1H₂O
MW: 92.05 g/mol
Melting Pt: 48…52 °C
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00127974
CAS Number: 563-96-2
EINECS: 206-058-5
UN: 3261
ADR: 8,II

Order Now

Specification Test Results

Appearance White to light yellow Crystalline powder and/or chunks
Infrared spectrum Conforms
Melting point 48 to 52 °C
Titration with NaOH ≥97.5 %
NMR (1H-NMR) Consistent with structure

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR