2,3-Dimercapto-1-propanol 97.0-100.5%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 2,3-DimercaptopropanolDithioglycerineDimercaprolDimercapto-2,3 propanol-12,3-Dimercaptopropane-1-ol2,3-Dimercaptopropan-1-olBAL

11530-0250 11530-0050
TS11530-0050EA 136 USD
TS11530-0050 TS11530-0250
2,3-Dimercapto-1-propanol 97.0-100.5%
2,3-Dimercapto-1-propanol
Formula: C₃H₈OS₂
MW: 124.22 g/mol
Boiling Pt: 120 °C (15 mmHg)
Density: 1.243 g/cm³
Flash Pt: >112 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00004864
CAS Number: 59-52-9
EINECS: 200-433-7
UN: 2810
ADR: 6.1,III

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5700 to 1.5750 (20 °C, 589 nm)
Titration Iodimetric 97.0 to 100.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR