2',4'-Dihydroxyacetophenone 98%

Supplier: Thermo Scientific Chemicals
Warning

11480-0050 11480-1000 11480-5000
TS11480-0050EA 31 USD
TS11480-0050 TS11480-1000 TS11480-5000
2',4'-Dihydroxyacetophenone 98%
2',4'-Dihydroxyacetophenone
Formula: C₈H₈O₃
Boiling Pt: 336 °C (1013 hPa)
Melting Pt: 143…147 °C
Density: 1.18 g/cm³
MDL Number: MFCD00002279
CAS Number: 89-84-9
EINECS: 201-945-3
Merck Index: 12,08308

Order Now

Specification Test Results

Appearance Off-white to pink to brown Crystalline powder or crystals
Infrared spectrum Conforms
Melting point 143 to 147 °C
HPLC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR