Citral cis- and trans-mixture 95%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
3,7-Dimethylocta-2,6-dienal
11044-5000
11044-0050
11044-1000
TS11044-5000EA
120
USD
TS11044-5000
TS11044-0050
TS11044-1000
Citral cis- and trans-mixture 95%
Citral
Formula:
C₁₀H₁₆O MW: 152.24 g/mol Boiling Pt: 229 °C (1013 hPa) Melting Pt: –10 °C Density: 0.889 g/cm³ Flash Pt: 101 °C Storage Temperature: Ambient |
MDL Number:
MFCD00006997 CAS Number: 5392-40-5 EINECS: 226-394-6 Merck Index: 13,02346 |
Specification Test Results
Appearance | Clear yellow Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4860 to 1.4900 (20 °C, 589 nm) |
Density | (20/4°C) 0.886 to 0.893 kg/l |
GC | ≥94 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...