1-Phenyl-1,3-butandione 98%

Supplier: Thermo Scientific Chemicals

Synonyms: Benzoylacetone1-Phenylbutane-1,3-dione1-Phenyl-1,3-butanedione1-Benzoylacetone

10569-2500 10569-0500
TS10569-2500EA 270 USD
TS10569-2500 TS10569-0500
1-Phenyl-1,3-butandione 98%
1-Phenyl-1,3-butandione
Formula: C₆H₅C(O)CH₂C(O)CH₃
MW: 162.19 g/mol
Melting Pt: 54…58 °C
Density: 1.09 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00008786
CAS Number: 93-91-4
EINECS: 202-286-4

Order Now

Specification Test Results

Appearance Yellow Crystalline powder or flakes
Infrared spectrum Conforms
Melting point 54 to 58 °C
GC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR