Dibac® Liquid bleach, sanitizer, deodorizer, cleaning additive, fogging sanitizer
Supplier: Diversey
08500153 08500330 08500470 08500280 08500100
76547-712GL 258747.41 USD
76547-712 76547-714 76547-710 76547-716 76547-718
Dibac® Liquid bleach, sanitizer, deodorizer, cleaning additive, fogging sanitizer
Cleaning Agents Disinfectants
Dibac® is a concentrated liquid sodium hypochlorite product, excellent for sanitizing and deodorizing food and beverage plants and equipment.
  • Liquid product: easy to feed
  • Effective sanitizer: passes A.O.A.C. Germicidal and Detergent Sanitizers test
  • Convenient: goes into use solution immediately