4'-Fluoro-2'-nitroacetanilide 99+%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: N-(4-Fluoro-2-nitrophenyl)acetamide

24966-0250
TS24966-0250EA 104 USD
TS24966-0250
4'-Fluoro-2'-nitroacetanilide 99+%
4'-Fluoro-2'-nitroacetanilide
Formula: C₈H₇FN₂O₃
MW: 179.22 g/mol
Melting Pt: 70…73 °C
Storage Temperature: Ambient
MDL Number: MFCD00031517
CAS Number: 448-39-5

Order Now

Specification Test Results

Appearance Ochre Crystalline powder
Infrared spectrum Conforms
Melting point 70 to 73 °C
GC ≥99.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR