1-Bromo-2-chloroethane 98%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Ethylene chlorobromide

21418-1000 21418-5000
TS21418-1000EA 62 USD
TS21418-1000 TS21418-5000
1-Bromo-2-chloroethane 98%
1-Bromo-2-chloroethane
Formula: BrH₂CCH₂Cl
Boiling Pt: 106…107 °C (760 mmHg)
Melting Pt: –17 °C
Density: 1.73 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00000962
CAS Number: 107-04-0
EINECS: 203-456-0
UN: 2810
ADR: 6.1,III

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4900 to 1.4920 (20°C, 589 nm)
GC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR