1-Iodobutane 98% stabilised
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
1-Butyl iodide
17343-1000
17343-5000
TS17343-1000EA
74
USD
TS17343-1000
TS17343-5000
1-Iodobutane 98% stabilised
1-Iodobutane
Formula:
C₄H₉I Boiling Pt: 130 °C (1013 hPa) Melting Pt: –103 °C Density: 1.61 g/cm³ Flash Pt: 33 °C Storage Temperature: Ambient |
MDL Number:
MFCD00001098 CAS Number: 542-69-8 EINECS: 208-824-4 UN: 2929 ADR: 6.1,II Merck Index: 13,01572 |
Specification Test Results
Appearance | Clear colorless to light yellow to light pink Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4975 to 1.4995 (20°C, 589 nm) |
GC | ≥97.5 % |
Stabilizer | copper |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...