1-Bromopentane 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
15252-0010
15252-2500
TS15252-0010EA
189
USD
TS15252-0010
TS15252-2500
1-Bromopentane 98%
1-Bromopentane
Formula:
BrH₂C(CH₂)₃CH₃ MW: 151.05 g/mol Boiling Pt: 129 Melting Pt: -95 Density: 1.21 g/cm³ Flash Pt: 79 °C Storage Temperature: Ambient |
MDL Number:
MFCD00000267 CAS Number: 110-53-2 EINECS: 203-776-0 UN: 1993 ADR: 3 III Merck Index: 13,00606 |
Specification Test Results
Appearance | Clear colorless to light yellow Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4430 to 1.4460 (20°C, 589 nm) |
GC | ≥97.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...