4-Toluoyl chloride 98%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: p-Toluoyl chloride, 4-Methylbenzene-1-carbonyl chloride, p-Methylbenzoyl chloride, 4-Methylbenzoyl chloride

14798-0010 14798-0050 14798-2500
TS14798-0010EA 237 USD
TS14798-0010 TS14798-0050 TS14798-2500
4-Toluoyl chloride 98%
4-Toluoyl chloride
Formula: C₈H₇ClO
MW: 154.6 g/mol
Boiling Pt: 225
Melting Pt: -2
Density: 1.16 g/cm³
Flash Pt: 82 °C
Storage Temperature: Ambient
MDL Number: MFCD00000696
CAS Number: 874-60-2
EINECS: 212-864-8
UN: 3265
ADR: 8 II

Order Now

Specification Test Results

Appearance Clear colorless to light brown Liquid
Infrared spectrum Conforms
Refractive index 1.5525 to 1.5545 (20°C, 589 nm)
Titration RCOCl ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR