n-Undecane 99%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Undecane

14066-0025 14066-5000 14066-0100 14066-1000
TS14066-0025EA 734 USD
TS14066-0025 TS14066-5000 TS14066-0100 TS14066-1000
n-Undecane 99%
n-Undecane
Formula: H₃C(CH₂)₉CH₃
MW: 156.31 g/mol
Boiling Pt: 196 °C (760 mmHg)
Melting Pt: –26 °C
Density: 0.74 g/cm³
Flash Pt: 65 °C
Storage Temperature: Ambient
MDL Number: MFCD00008959
CAS Number: 1120-21-4
EINECS: 214-300-6
UN: 2330
ADR: 3,III

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.4160 to 1.4180 (20°C, 589 nm)
GC ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR