2,2,2-Trichloroethanol 99%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
2,2,2-Trichloroethan-1-ol
13944-1000
13944-5000
TS13944-1000EA
55
USD
TS13944-1000
TS13944-5000
2,2,2-Trichloroethanol 99%
2,2,2-Trichloroethanol
Formula:
C₂H₃Cl₃O Boiling Pt: 151 °C (1013 hPa) Melting Pt: 17.8 °C Density: 1.55 g/cm³ Flash Pt: 88 °C Storage Temperature: Ambient |
MDL Number:
MFCD00004677 CAS Number: 115-20-8 EINECS: 204-071-0 Merck Index: 12,09786 |
Specification Test Results
Appearance | Clear colorless to light yellow Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4880 to 1.4910 (20°C, 589 nm) |
GC | ≥98.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...