α,α,α',α'-Tetrabromo-o-xylene 97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
1,2-Bis(dibromomethyl)benzene
13797-0250
13797-1000
TS13797-0250EA
43
USD
TS13797-0250
TS13797-1000
α,α,α',α'-Tetrabromo-o-xylene 97%
α,α,α',α'-Tetrabromo-o-xylene
Formula:
C₈H₆Br₄ Boiling Pt: 346 °C (1013 hPa) Melting Pt: 114…118 °C Storage Temperature: Ambient |
MDL Number:
MFCD00000131 CAS Number: 13209-15-9 EINECS: 236-176-2 UN: 3261 ADR: 8,II |
Specification Test Results
Appearance | Off-white to beige to light grey Crystalline powder |
Infrared spectrum | Conforms |
Melting point | 114 to 118 °C |
GC | ≥96.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...