Chlorocyclohexane 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Cyclohexyl chloride
11130-0010
11130-1000
11130-2500
TS11130-0010EA
114
USD
TS11130-0010
TS11130-1000
TS11130-2500
Chlorocyclohexane 98%
Chlorocyclohexane
Formula:
C₆H₁₁Cl Boiling Pt: 142 °C (1013 hPa) Melting Pt: –44 °C Density: 1 g/cm³ Flash Pt: 28 °C Storage Temperature: Ambient |
MDL Number:
MFCD00003822 CAS Number: 542-18-7 EINECS: 208-806-6 UN: 1993 ADR: 3,III Merck Index: 13,02761 |
Specification Test Results
Appearance | Clear liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4610 to 1.4630 (20°C, 589 nm) |
Water | ≤0.05 % |
GC | ≥97.5 % |
Color scale | ≤10 APHA |
Water soluble titr. acid | ≤0.0005 meq/g |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...