Coumarin 99+%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 1-Benzopyran-2-one, 2H-Chromen-2-one, 1,2-Benzopyrone

11053-0010 11053-0050 11053-2500
TS11053-0010EA 90 USD
TS11053-0010 TS11053-0050 TS11053-2500
Coumarin 99+%
Coumarin
Formula: C₉H₆O₂
MW: 146.15 g/mol
Boiling Pt: 298 °C (1013 hPa)
Melting Pt: 68…71 °C
Flash Pt: 162 °C
Storage Temperature: Ambient
MDL Number: MFCD00006850
CAS Number: 91-64-5
EINECS: 202-086-7
UN: 2811
ADR: 6.1,III
Merck Index: 13,02588

Order Now

Specification Test Results

Appearance White Crystalline powder or crystals
Infrared spectrum Conforms
Melting point 68 to 71 °C
GC ≥99.0 %
Specific gravity (20 °C) 0.868 to 0.888
Acid value ≤1 mg KOH/g

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR