CloneDetect, Molecular Devices
Supplier: Molecular Devices
K8200 K8205 K8225
76411-576EA 475.1 USD
76411-576 76411-568 76411-562
CloneDetect, Molecular Devices
Assays Cellular Assays
Screen and select high value mammalian clones with label-free, in situ detection of IgG secretion with the ClonePix system, and other cell analyzers with a green fluorescent channel.
  • Screen and select high-value mammalian clones with label-free, in situ detection of IgG secretion
  • A choice of detection agents specific for secreted mouse, rat or human IgGs
  • Sterile, virus and mycoplasma free
  • Optimized for ClonePix system, and compatible with cell analyzers with a green fluorescent channel
A cell-culture validated, sterile and azide-free antibody reagent for detecting IgG in mammalian cell clone screening.