Castor oil ethoxylated pH 6.0-8.1
Supplier: Thermo Scientific Chemicals
39728-0010
39728-2500
TS39728-0010EA
154
USD
TS39728-0010
TS39728-2500
Castor oil ethoxylated pH 6.0-8.1
Castor oil ethoxylated
Boiling Pt:
>100 °C (760 mmHg) Melting Pt: 4 °C Density: 1.05 g/cm³ Flash Pt: 257 °C Storage Temperature: Ambient |
MDL Number:
MFCD00130879 CAS Number: 61791-12-6 |
Specification Test Results
Appearance | White to yellow Viscous liquid |
Infrared spectrum | Conforms |
Water | ≤3.0 % |
pH | 6.0 to 8.0 (10 % in water) |
Hydroxyl value | 65 to 78 mg KOH/g |
Iodine value | 25 to 35 g I2/100 g |
Saponification value | 65 to 70 mg KOH/g |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...