1,3-benzothiazol-5-amine 95%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 1,3-Benzothiazol-5-amine, 5-Aminobenzo[d]thiazole, 1,3-Benzothiazol-5-amine, 5-Aminobenzothiazole

45154-0050 45154-0250
TS45154-0050EA 522 USD
TS45154-0050 TS45154-0250
1,3-benzothiazol-5-amine 95%
1,3-benzothiazol-5-amine
Formula: C₇H₆N₂S
Melting Pt: 73…77.5 °C
Storage Temperature: Ambient
MDL Number: MFCD04115282
CAS Number: 1123-93-9

Order Now

Specification Test Results

Appearance White to yellow Powder or solid
Infrared spectrum Conforms
HPLC ≥94.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR