Sodium octanoate 98%

Supplier: Thermo Scientific Chemicals
Warning

33460-0025 33460-1000 33460-5000
TS33460-0025EA 794 USD
TS33460-0025 TS33460-1000 TS33460-5000
Sodium octanoate 98%
Sodium octanoate
Formula: C₈H₁₅NaO₂
Melting Pt: >225 °C
Storage Temperature: Ambient
MDL Number: MFCD00058511
CAS Number: 1984-06-1
EINECS: 217-850-5

Order Now

Specification Test Results

Appearance White to off-white Powder
Infrared spectrum Conforms
Water ≤5 % (K.F.)
Titration with HClO4 ≥97.5 % (on dry substance)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR