Ethanesulphonyl chloride 98+%

Supplier: Thermo Scientific Chemicals
Danger

21582-0025 21582-0050 21582-0500 21582-1000 21582-5000
TS21582-0025EA 720 USD
TS21582-0025 TS21582-0050 TS21582-0500 TS21582-1000 TS21582-5000
Ethanesulphonyl chloride 98+%
Ethanesulphonyl chloride
Formula: C₂H₅ClO₂S
MW: 128.58 g/mol
Boiling Pt: 177
Melting Pt: -68
Density: 1.35 g/cm³
Flash Pt: 83 °C
Storage Temperature: Ambient
MDL Number: MFCD00007460
CAS Number: 594-44-5
EINECS: 209-842-5
UN: 2927
ADR: 6.1 II

Order Now

Specification Test Results

Appearance Clear colorless to yellow to light brown Liquid (May darken on storage)
Infrared spectrum Conforms
Refractive index 1.4520 to 1.4540 (20°C, 589 nm)
GC ≥98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR