Tetraphenylphosphonium bromide 99%

Supplier: Thermo Scientific Chemicals
Warning

20402-0050 20402-0250 20402-1000
TS20402-0050EA 28 USD
TS20402-0050 TS20402-0250 TS20402-1000
Tetraphenylphosphonium bromide 99%
Tetraphenylphosphonium bromide
Formula: C₂₄H₂₀BrP
Melting Pt: 295…299 °C
Flash Pt: 260 °C
Storage Temperature: Ambient
MDL Number: MFCD00011915
CAS Number: 2751-90-8
EINECS: 220-393-4

Order Now

Specification Test Results

Appearance White to off-white Crystalline powder or crystals
Infrared spectrum Conforms
Melting point 296 to 300 °C
Titration Argentometric ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR