Bromocyclobutane 97%, pure
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Cyclobutyl bromide
32050-0010
32050-0050
32050-0250
TS32050-0010EA
69
USD
TS32050-0010
TS32050-0050
TS32050-0250
Bromocyclobutane 97%, pure
Bromocyclobutane
Formula:
C₄H₇Br MW: 135 g/mol Boiling Pt: 108 °C Density: 1.43 g/cm³ Flash Pt: 22 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00001317 CAS Number: 4399-47-7 EINECS: 224-530-9 UN: 1993 ADR: 3,II |
Specification Test Results
Appearance | Clear colorless to yellow Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4780 to 1.4810 (20°C, 589 nm) |
GC | ≥96.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...