trans-1,4-Dibromo-2-butene 97%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: (E)-1,4-Dibromobut-2-ene, (E)-1,4-Dibromo-2-butene

40646-0250 40646-1000 40646-5000
TS40646-0250EA 92 USD
TS40646-0250 TS40646-1000 TS40646-5000
trans-1,4-Dibromo-2-butene 97%
trans-1,4-Dibromo-2-butene
Formula: C₄H₆Br₂
MW: 213.9
Boiling Pt: 203
Melting Pt: 48…53 °C
Flash Pt: 113 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00000249
CAS Number: 821-06-7
EINECS: 212-472-7
UN: 2923
ADR: 8 II

Order Now

Specification Test Results

Appearance White to beige to brown Crystalline powder or crystals
Infrared spectrum Conforms
Melting point 48 to 53 °C
GC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR