Submersible Pumps for Corrosive/Acid Sumps
Supplier: BPH PUMP & EQUIPMENT INC
GF32-9 GF32-9NL IGF32-9 IGF32-FNLZ-115 TIGF32-9NL
MFLX75500-80EA 1650 USD
MFLX75500-80 MFLX75500-82 MFLX75500-84 MFLX75500-86 MFLX75500-88
Submersible Pumps for Corrosive/Acid Sumps
Pumps Centrifugal Pumps
Only electric submersible pump with entire motor encapsulated in Noryl®.
  • Noryl material never rusts and provides excellent resistance against acids and alkalis
  • Compact and lightweight—only 13-1⁄2 lbs—with convenient carrying handle
  • Virtually unbreakable
These pumps can easily be installed in a sump and are ideal for safeguarding vaults, emptying tanks, or pumping out ponds. The unique dual-port design virtually eliminates kinks in your hose; use vertical port for deep/narrow places or use the horizontal port for shallow/wide applications. Thermal breaker incorporated in motor windings shuts off power before over-heating occurs due to locked rotor or long periods of running dry.
For permanent installation, temperature should not exceed 135 °F (57 °C). For intermittent use, submerge less than 20 minutes in 195 °F (90 °C) maximum.