Big Blue General-Purpose Pleated Filter Cartridges
Supplier: Big Blue
255494-43 155120-43 155405-43 155305-43
MFLX01508-86EA 38.4 USD
MFLX01508-86 MFLX01512-88 MFLX01512-76 MFLX01512-74
Big Blue General-Purpose Pleated Filter Cartridges
Filters Filter Cartridges
Design offers higher flow rates and dirt-loading capacities than spun or wound depth cartridges, especially when removing lower micron-sized particles.
  • Remove non-gelatinous particles that fall into a narrow size range, typically in post filtration or polishing applications
Pleated cartridges separate viscous sediments and flat particulates. Use polyester cartridges for water filtration and polypropylene cartridges with harsher chemicals. Plastisol end caps.
Note: Cartridges are for sediment filtration only; they will not remove bacteria.