Economical Carbon-Impregnated Paper Cartridge
Supplier: PENTAIR RESIDENTIAL FILTRATION LLC
155597-43 155002-43
MFLX01509-25EA 19.7 USD
MFLX01509-25 MFLX01509-27
Economical Carbon-Impregnated Paper Cartridge
Filters Filter Cartridges
Ideal for chemically neutral waters with lower chlorine concentrations.
  • Constructed of carbon-impregnated cellulose
This cellulose cartridge removes organic tastes/odors and chlorine, plus reduces sediment. Suitable to 125 °F (52 °C).