Methanesulphonamide 98%

Supplier: Thermo Scientific Chemicals
Warning

29611-0250 29611-1000
TS29611-0250EA 95 USD
TS29611-0250 TS29611-1000
Methanesulphonamide 98%
Methanesulphonamide
Formula: CH₅NO₂S
Melting Pt: 88…92 °C
Storage Temperature: Ambient
MDL Number: MFCD00007940
CAS Number: 3144-09-0
EINECS: 221-553-6

Order Now

Specification Test Results

Appearance White to yellow Powder or crystals or flakes
Infrared spectrum Conforms
Melting point 88 to 92 °C
GC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR