Cyanoacetic acid 97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
11067-0010
11067-0050
11067-0250
11067-2500
TS11067-0010EA
119
USD
TS11067-0010
TS11067-0050
TS11067-0250
TS11067-2500
Cyanoacetic acid 97%
Cyanoacetic acid
Formula:
C₃H₃NO₂ Boiling Pt: 108 °C (15 mmHg) Melting Pt: 65…67 °C Flash Pt: 107 °C Storage Temperature: Ambient |
MDL Number:
MFCD00002677 CAS Number: 372-09-8 EINECS: 206-743-9 UN: 3261 ADR: 8,II Merck Index: 13,02718 |
Specification Test Results
Appearance | White to light beige Adhering crystalline powder or crystals |
Infrared spectrum | Conforms |
Titration with NaOH | ≥96.0 % |
Specific optical rotation | -10° to -12° (20°C, 589 nm) (neat) |
Titration with HClO4 | ≥97.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...