5-Methylphenazinium methyl sulphate 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Phenazine methosulfate, PMS, 5-Methylphenazin-5-ium methyl sulfate, 5-Methylphenazin-5-ium methyl sulphate, Phenazine methosulphate, Phenazine methylsulfate, Phenazine methylsulphate

13016-0010 13016-0100 13016-0250
TS13016-0010EA 56 USD
TS13016-0010 TS13016-0100 TS13016-0250
5-Methylphenazinium methyl sulphate 98%
5-Methylphenazinium methyl sulphate
Formula: C₁₄H₁₄N₂O₄S
Melting Pt: 158…160 °C
Storage Temperature: Freezer
MDL Number: MFCD00011923
CAS Number: 299-11-6
EINECS: 206-072-1
Merck Index: 12,06185

Order Now

Specification Test Results

Appearance Dark yellow to khaki to brown Crystalline powder
Infrared spectrum Conforms
UV spectrophotometry ≥97.5 % (0.01g/l H2O, 1cm cell, lambda max 387.5 nm)
GC ≥96.0 %
UV at 260 nm A: ≤0.02
Titration after Ion exchange ≥99.0 % (on dry substance)
Membrane filtration test (on 0.4 M soln.) Passes test
Suitability (HPLC gradient) Passes test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR