4-Hydroxy-3-methoxycinnamic acid (Ferulic acid) 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 3-(4-Hydroxy-3-methoxyphenyl)acrylic acid, NSC 2821, NSC 674320, NSC 51986, Coniferic acid, Ferulic acid

15636-0050 15636-0250 15636-1000
TS15636-0050EA 32 USD
TS15636-0050 TS15636-0250 TS15636-1000
4-Hydroxy-3-methoxycinnamic acid (Ferulic acid) 99%
4-Hydroxy-3-methoxycinnamic acid (Ferulic acid)
Formula: C₁₀H₁₀O₄
Melting Pt: 169…173 °C
MDL Number: MFCD00004400
CAS Number: 1135-24-6
EINECS: 208-679-7

Order Now

Specification Test Results

Appearance Off-white to beige Powder
Infrared spectrum Conforms
Melting point 169 to 173 °C
HPLC ≥98.5 %
Titration with NaOH ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR