2-Aminoethyl hydrogen sulfate 99%

Supplier: Thermo Scientific Chemicals

Synonyms: 2-Aminoethyl Hydrogen sulfate, Sulfuric acid Mono(2-aminoethyl) Ester, 2-Aminoethyl Hydrogen sulphate

10364-1000 10364-5000
TS10364-1000EA 83 USD
TS10364-1000 TS10364-5000
2-Aminoethyl hydrogen sulfate 99%
2-Aminoethyl hydrogen sulphate
Formula: C₂H₇NO₄S
Melting Pt: 277…280 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00008179
CAS Number: 926-39-6
EINECS: 213-135-7

Order Now

Specification Test Results

Appearance White to light beige Crystalline powder
Infrared spectrum Conforms
Titration with NaOH ≥98.5 %
Clarity of solution Passes test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR