Methylenediphosphonic acid 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
21507-0010
21507-0050
TS21507-0010EA
86
USD
TS21507-0010
TS21507-0050
Methylenediphosphonic acid 98%
Methylenediphosphonic acid
Formula:
CH₆O₆P₂ Boiling Pt: 82 °C (6 mmHg) Melting Pt: 200…204 °C Storage Temperature: Ambient |
MDL Number:
MFCD00002139 CAS Number: 1984-15-2 EINECS: 217-851-0 UN: 3261 ADR: 8,II |
Specification Test Results
Appearance | White Crystalline powder |
Infrared spectrum | Conforms |
Melting point | 197 to 204 °C |
Titration with NaOH | ≥97.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...