Isopropyl myristate 96%
Supplier: Thermo Scientific Chemicals
Synonyms:
Isopropyl tetradecanoate
, Tetradecylic acid isopropyl ester
, Myristic acid isopropyl ester
, Tetradecanoic acid isopropyl ester
41285-1000
41285-0250
41285-0010
TS41285-0010EA
178
USD
TS41285-0010
TS41285-1000
TS41285-0250
Isopropyl myristate 96%
Isopropyl myristate
Formula:
C₁₇H₃₄O₂ MW: 270.46 g/mol Boiling Pt: 167 °C (9 mmHg) Density: 0.85 g/cm³ Flash Pt: >110 °C Storage Temperature: Ambient |
MDL Number:
MFCD00008982 CAS Number: 110-27-0 EINECS: 203-751-4 Merck Index: 12,05234 |
Specification Test Results
Appearance | Clear liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4340 to 1.4360 (20°C, 589 nm) |
Water | ≤0.1 % |
GC | ≥95 % |
Color scale | ≤30 APHA |
Acid value | ≤0.5 mg KOH/g |
Iodine value | ≤1 g I2/100 g |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...