Biphenyl (PCB No. 0) 99%

Supplier: Thermo Scientific Chemicals
Warning

10625-0010 10625-0050 10625-2500
TS10625-0010EA 47 USD
TS10625-0010 TS10625-0050 TS10625-2500
Biphenyl (PCB No. 0) 99%
Biphenyl (PCB No. 0)
Formula: C₁₂H₁₀
Boiling Pt: 255 °C (760 mmHg)
Melting Pt: 68…72 °C
Flash Pt: 113 °C
Storage Temperature: Ambient
MDL Number: MFCD00003054
CAS Number: 92-52-4
EINECS: 202-163-5
UN: 3077
ADR: 9,III
Merck Index: 13,03346

Order Now

Specification Test Results

Appearance White to light yellow Crystalline powder or crystals or flakes
Infrared spectrum Conforms
Melting point 68 to 72 °C
GC ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR