Trimethyl phosphate 99%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Trimethylphosphate

15797-0025 15797-0500 15797-5000
TS15797-0025EA 273 USD
TS15797-0025 TS15797-0500 TS15797-5000
Trimethyl phosphate 99%
Trimethyl phosphate
Formula: C₃H₉O₄P
MW: 140.08 g/mol
Boiling Pt: 197.2
Melting Pt: -46
Density: 1.197 g/cm³
Flash Pt: 107 °C
Storage Temperature: Ambient
MDL Number: MFCD00008348
CAS Number: 512-56-1
EINECS: 208-144-8

Order Now

Specification Test Results

Appearance Clear liquid
Infrared spectrum Conforms
Refractive index 1.3930 to 1.3970 (20°C, 589 nm)
Water ≤0.2 %
GC ≥98.5 %
Color scale ≤30 APHA
Residue after evaporation ≤0.001 %
Acidity ≤0.0005 meq/g
Alkalinity ≤0.0002 meq/g
Aluminium (Al) ≤0.5 ppm
Barium (Ba) ≤0.1 ppm
Boron (B) ≤0.02 ppm
Cadmium (Cd) ≤0.05 ppm
Calcium (Ca) ≤0.5 ppm
Chromium (Cr) ≤0.02 ppm
Cobalt (Co) ≤0.02 ppm
Copper (Cu) ≤0.02 ppm
Iron (Fe) ≤0.1 ppm
Lead (Pb) ≤0.1 ppm
Magnesium (Mg) ≤0.1 ppm
Manganese (Mn) ≤0.02 ppm
Nickel (Ni) ≤0.02 ppm
Tin (Sn) ≤0.1 ppm
Zinc (Zn) ≤0.1 ppm
Carbonyl compounds ≤0.01 % (as butyraldehyde)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR