di-Sodium L(+)-tartrate dihydrate 99+% ACS

Supplier: Thermo Scientific Chemicals

42444-0025 42444-0100 42444-1000 42444-5000
TS42444-0025EA 565 USD
TS42444-0025 TS42444-0100 TS42444-1000 TS42444-5000
di-Sodium L(+)-tartrate dihydrate 99+% ACS
di-Sodium L(+)-tartrate dihydrate
Formula: NaOOC(CHOH)₂COONa·2H₂O
Storage Temperature: Ambient
MDL Number: MFCD00150035
CAS Number: 6106-24-7
EINECS: 212-773-3
Merck Index: 12,08833
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=39a75038f4ff5e6c234267b15540823ae361bed4&vid=89289572c48b0c6415f8f11e916a51f66e129474&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White Powder or needles
Infrared spectrum Conforms
Loss on drying 15.61 % to 15.71 % (150°C)
pH 7.0 to 9.0 (5 % at 25°C)
Titration with HClO4 99.0 to 101.0 %
Chloride (Cl) ≤5 ppm
Phosphate (PO4) ≤5 ppm
Sulfate (SO4) ≤50 ppm
Ammonium (NH4) ≤30 ppm
Calcium (Ca) ≤100 ppm
Iron (Fe) ≤10 ppm
Heavy metals (as Pb) ≤5 ppm
Insoluble matter ≤0.005 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR