Oleic acid 97%
Supplier: Thermo Scientific Chemicals
Synonyms:
(9Z)-9-Octadecenoic acid
27029-0250
27029-0050
27029-1000
TS27029-0050EA
81.2
USD
TS27029-0050
TS27029-0250
TS27029-1000
Oleic acid 97%
Oleic acid
Formula:
C₁₈H₃₄O₂ MW: 282.47 g/mol Boiling Pt: 360 °C (760 mmHg) Melting Pt: 13 °C Density: 0.89 g/cm³ Flash Pt: 189 °C Storage Temperature: Freezer |
MDL Number:
MFCD00064242 CAS Number: 112-80-1 EINECS: 204-007-1 Merck Index: 13,06898 |
Specification Test Results
Appearance | Clear colorless to light yellow Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4580 to 1.4610 (20°C, 589 nm) |
GC | ≥96.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...