Dehydroacetic acid 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 3-Acetyl-6-methyl-2H-pyran-2,4(3H)-dione3-Acetyl-4-hydroxy-6-methyl-2H-pyran-2-one

11194-5000 11194-0050 11194-1000
TS11194-5000EA 174 USD
TS11194-5000 TS11194-0050 TS11194-1000
Dehydroacetic acid 98%
Dehydracetic acid
Formula: C₈H₈O₄
MW: 168.15 g/mol
Boiling Pt: 333 °C (1013 hPa)
Melting Pt: 109…112 °C
Storage Temperature: Ambient
MDL Number: MFCD00066709
CAS Number: 520-45-6
EINECS: 208-293-9
Merck Index: 13,02885

Order Now

Specification Test Results

Appearance Cream to light yellow Crystalline powder
Infrared spectrum Conforms
Melting point 109 to 112 °C
Sulfated ash ≤0.1 % (700 °C, 2 h)
Titration with NaOH ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR