1-Methylnaphthalene 96%
Supplier: Thermo Scientific Chemicals
|
Warning
|
12716-0025
12716-1000
12716-5000
TS12716-0025EA
465
USD
TS12716-0025
TS12716-1000
TS12716-5000
1-Methylnaphthalene 96%
1-Methylnaphthalene
Formula:
C₁₁H₁₀ Boiling Pt: 240…243 °C (760 mmHg) Melting Pt: –22 °C Density: 1.02 g/cm³ Flash Pt: 82 °C Storage Temperature: Ambient |
MDL Number:
MFCD00004034 CAS Number: 90-12-0 EINECS: 201-966-8 UN: 3082 ADR: 9,III |
Specification Test Results
Appearance | Clear colorless to yellow to light brown Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.6100 to 1.6180 (20°C, 589 nm) |
GC | ≥99.4 % |
Color scale | ≤10 APHA |
Aldehyde (C3H6O) | ≤0.01 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...