Trimesic acid 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Benzene-1,3,5-tricarboxylic acid, 1,3,5-Benzenetricarboxylic acid

10535-0050 10535-0500 10535-5000
TS10535-0050EA 31 USD
TS10535-0050 TS10535-0500 TS10535-5000
Trimesic acid 98%
Trimesic acid
Formula: C₉H₆O₆
MW: 210.14 g/mol
Melting Pt: 380 °C
Flash Pt: >250 °C
Storage Temperature: Ambient
MDL Number: MFCD00002517
CAS Number: 554-95-0
EINECS: 209-077-7

Order Now

Specification Test Results

Appearance White to beige Powder
Infrared spectrum Conforms
HPLC ≥97.5 %
UV Lambda max 590 to 596 nm (in Methanol)
Molar absorption coefficient 67400 l/(mol.cm) (at Lambda max)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR