1-Octadecanethiol 96%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
n-Octadecyl mercaptan, n-Octadecanethiol, Stearyl mercaptane, 1-Mercaptooctadecane, Octadecane-1-thiol
35498-0250
35498-1000
35498-5000
TS35498-0250EA
37
USD
TS35498-0250
TS35498-1000
TS35498-5000
1-Octadecanethiol 96%
1-Octadecanethiol
Formula:
C₁₈H₃₈S Boiling Pt: 366 °C (760 mmHg) Melting Pt: 25…35 °C Flash Pt: 185 °C Storage Temperature: Ambient |
MDL Number:
MFCD00004886 CAS Number: 2885-00-9 EINECS: 220-744-1 UN: 3335 ADR: 9,III |
Specification Test Results
Appearance | White Crystalline solid |
Infrared spectrum | Conforms |
Melting point | 25 to 35 °C |
GC | ≥95 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...