Formaldehyde 37% (w/w) in water stabilized

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: MethylaldehydeMethylaldehyde solutionFormolMethanal solutionFormalinFormaline solutionParaformaldehyde aqueous solution

Stabilization: Stabilized with methanol 5-15%

11969-0010
TS11969-0010EA 58.18 USD
TS11969-0010
Formaldehyde 37% (w/w) in water stabilized
Formaldehyde
Formula: CH₂O
MW: 30.03 g/mol
Melting Pt: –15 °C
Density: 1.083 g/cm³
Flash Pt: 50 °C
Storage Temperature: Ambient
MDL Number: MFCD00003274
CAS Number: 50-00-0
EINECS: 200-001-8
UN: 1198
ADR: 3,III

Order Now

Specification Test Results

Appearance Clear solution
Color scale ≤10 APHA
Stabilizer 5 to 15 % methanol
Titration 35.0 to 39.0 %
Free acid (HCOOH) ≤0.1 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR