Apixaban

Supplier: Thermo Scientific Chemicals
Danger

46449-1000 46449-5000
TS46449-1000EA 95 USD
TS46449-1000 TS46449-5000
Apixaban
Apixaban
Formula: C₂₅H₂₅N₅O₄
Storage Temperature: Freezer
MDL Number: MFCD11977295
CAS Number: 503612-47-3

Order Now

Specification Test Results

Appearance White to light yellow Crystalline powder
Infrared spectrum Conforms
Melting point 238 to 240 °C
HPLC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR