Valinomycin 98%

Supplier: Thermo Scientific Chemicals
Danger

38512-0100 38512-0500
TS38512-0100EA 96 USD
TS38512-0100 TS38512-0500
Valinomycin 98%
Valinomycin
Formula: C₅₄H₉₀O₁₈N₆
MW: 1111.34 g/mol
Melting Pt: 190 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00005114
CAS Number: 2001-95-8
UN: 2811
ADR: 6.1 I

Order Now

Specification Test Results

Appearance White Powder
Melting point 186 to 191°C
Solubility (50mg/ml CH2Cl2) Clear colorless solution
HPLC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR