Maltotriose 93%

Supplier: Thermo Scientific Chemicals

22594-0250 22594-5000
TS22594-0250EA 651 USD
TS22594-0250 TS22594-5000
Maltotriose 93%
Maltotriose
Formula: C₁₈H₃₂O₁₆
Melting Pt: 132…135 °C
Storage Temperature: Ambient
MDL Number: MFCD00006629
CAS Number: 1109-28-0

Order Now

Specification Test Results

Appearance White Crystalline powder or crystals
Infrared spectrum Conforms
Water ≤5 % (K.F.)
HPLC ≥91 % (ELSD)
Specific optical rotation +160° to +165° (25°C, 589 nm) (c=1, H2O)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR