(±)-Propranolol hydrochloride 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 1-(Isopropylamino)-3-(1-naphthyloxy)-2-propanol hydrochloride, Propranolol hydrochloride, DL-Propranolol hydrochloride, (±)-1-Isopropylamino-3-(1-naphthyloxy)-2-propanol hydrochloride

20732-0050 20732-0250 20732-1000
TS20732-0050EA 160 USD
TS20732-0050 TS20732-0250 TS20732-1000
(±)-Propranolol hydrochloride 99%
(±)-Propranolol hydrochloride
Formula: C₁₆H₂₁NO₂·HCl
MW: 295.81 g/mol
Melting Pt: 163…166 °C
Storage Temperature: Ambient
MDL Number: MFCD00012558
CAS Number: 318-98-9

Order Now

Specification Test Results

Appearance White to light yellow Powder
Infrared spectrum Conforms
Melting point 163 to 166 °C
Titration with HClO4 ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR