Potassium oxonate 97.5%

Supplier: Thermo Scientific Chemicals

16853-0250 16853-1000
TS16853-0250EA 131 USD
TS16853-0250 TS16853-1000
Potassium oxonate 97.5%
Potassium oxonate
Formula: C₄H₃N₃O₄
Melting Pt: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD00010565
CAS Number: 2207-75-2

Order Now

Specification Test Results

Appearance White Crystalline powder
Infrared spectrum Conforms
Titration with HClO4 ≥96.50 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR