Pectin pract.
Supplier: Thermo Scientific Chemicals
41686-0010
41686-0025
41686-2500
TS41686-0010EA
251
USD
TS41686-0010
TS41686-0025
TS41686-2500
Pectin pract.
Pectin
Storage Temperature:
Ambient |
MDL Number:
MFCD00081838 CAS Number: 9000-69-5 |
Specification Test Results
Appearance | Off-white to beige to golden Powder |
Assay | 67 to 71 % (degree of esterification) |
Loss on drying | ≤12 % (105°C, 2 hrs) |
Particle size | ≤2 % (> 60 mesh) |
Viscosity | 30.5 to 36.5 |
pH | 3.0 to 3.8 (1 % solution) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...